missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonyl fluoride, 98%

Catalog No. p-7045845
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1220414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1220414 Supplier Thermo Scientific Chemicals Supplier No. L1220414

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 455-16-3 | C7H7FO2S | 174.189 g/mol

p-Toluenesulfonyl fluoride is used for sulfonylation of organometallic reagents as a simple, one-step synthesis of sulfones. It is useful in the treatment of Alzheimer?s disease. It is used to study the DNS polymerase from spinach leaf extracts. It was used in isolation and separation of Pseudomonas aeruginosa membranes by density sucrose gradient centrifugation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Incompatible with bases, moisture and oxidizing agents. Store under dry inert gas. Protect from humidity and water.

Chemical Identifiers

CAS 455-16-3
Molecular Formula C7H7FO2S
Molecular Weight (g/mol) 174.189
MDL Number MFCD00007421
InChI Key IZZYABADQVQHLC-UHFFFAOYSA-N
Synonym p-toluenesulfonyl fluoride, tosyl fluoride, 4-toluenesulfonyl fluoride, benzenesulfonyl fluoride, 4-methyl, 4-tolylsulfonyl fluoride, toluene-4-sulfonyl fluoride, p-methylbenzenesulfonyl fluoride, p-toluenesulphonyl fluoride, toluene-p-sulphonyl fluoride, p-methylphenylsulfonyl fluoride
PubChem CID 9965
IUPAC Name 4-methylbenzenesulfonyl fluoride
SMILES CC1=CC=C(C=C1)S(=O)(=O)F

Specifications

Melting Point 40°C to 41°C
Boiling Point 112°C (16 mmHg)
Flash Point 105°C (221°F)
Quantity 25 g
UN Number UN3261
Beilstein 2208423
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 174.19
Percent Purity 98%
Chemical Name or Material p-Toluenesulfonyl fluoride
Show More Show Less
Hazard Category H302+H312+H332-H314
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 207-238-6
RTECSNumber XT8050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only