missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonyl Chloride 99.0+%, TCI America™

Catalog No. p-7136181
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T027225G 25 g
T0272500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T027225G Supplier TCI America Supplier No. T027225G
Only null left

Chemical Identifiers

CAS 98-59-9
Molecular Formula C7H7ClO2S
Molecular Weight (g/mol) 190.64
MDL Number MFCD00007450
InChI Key YYROPELSRYBVMQ-UHFFFAOYSA-N
Synonym tosyl chloride, p-toluenesulfonyl chloride, p-tosyl chloride, 4-toluenesulfonyl chloride, p-toluenesulfochloride, 4-methylbenzene-1-sulfonyl chloride, p-tolylsulfonyl chloride, p-toluenesulphonyl chloride, tosylchloride, p-toluenesulfonic acid chloride
PubChem CID 7397
IUPAC Name 4-methylbenzene-1-sulfonyl chloride
SMILES CC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 68°C
Color White
Boiling Point 146°C
Quantity 25 g
UN Number 3261
Formula Weight 190.64
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material p-Toluenesulfonyl Chloride
EINECSNumber (3)-1936
TSCA Yes