missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Toluenesulfonic Anhydride 95.0+%, TCI America™
Supplier: TCI America T148525G
Specifications
| p-Toluenesulfonic Anhydride | |
| 125°C | |
| C14H14O5S2 | |
| 25 g | |
| p-toluenesulfonic anhydride, 4-methylbenzenesulfonic anhydride, 4-toluenesulfonic anhydride, p-toluenesulfonicanhydride, p-toluene sulfonic anhydride, toluene-p-sulphonic anhydride, 4-methylphenyl sulfonyl 4-methylbenzenesulfonate, p-toluenesulphonic anhydride, toluene-4-sulfonic anhydride, benzenesulfonic acid, 4-methyl-, anhydride | |
| CC1=CC=C(C=C1)S(=O)(=O)OS(=O)(=O)C2=CC=C(C=C2)C | |
| 326.381 | |
| 326.38 | |
| Crystalline Powder |
| 4124-41-8 | |
| White | |
| MFCD00008548 | |
| 1759 | |
| PDVFSPNIEOYOQL-UHFFFAOYSA-N | |
| (4-methylphenyl)sulfonyl 4-methylbenzenesulfonate | |
| 77773 | |
| ≥95.0% (T) |
Chemical Identifiers
| 4124-41-8 | |
| 326.381 | |
| PDVFSPNIEOYOQL-UHFFFAOYSA-N | |
| 77773 | |
| CC1=CC=C(C=C1)S(=O)(=O)OS(=O)(=O)C2=CC=C(C=C2)C |
| C14H14O5S2 | |
| MFCD00008548 | |
| p-toluenesulfonic anhydride, 4-methylbenzenesulfonic anhydride, 4-toluenesulfonic anhydride, p-toluenesulfonicanhydride, p-toluene sulfonic anhydride, toluene-p-sulphonic anhydride, 4-methylphenyl sulfonyl 4-methylbenzenesulfonate, p-toluenesulphonic anhydride, toluene-4-sulfonic anhydride, benzenesulfonic acid, 4-methyl-, anhydride | |
| (4-methylphenyl)sulfonyl 4-methylbenzenesulfonate |
Safety and Handling
TSCA : No