Learn More
p-Toluenesulfinic acid, sodium salt, 97%, anhydrous, Thermo Scientific™
$89.63 - $257.90
Chemical Identifiers
| CAS | 824-79-3 |
|---|---|
| Molecular Formula | C7H7O2S·Na |
| Molecular Weight (g/mol) | 178.18 |
| InChI Key | KFZUDNZQQCWGKF-UHFFFAOYSA-M |
| Synonym | sodium 4-methylbenzenesulfinate, sodium p-toluenesulfinate, p-toluenesulfinic acid sodium salt, sodium toluene-4-sulphinate, sodium p-tolylsulfinate, sodium 4-toluenesulfinate, benzenesulfinic acid, 4-methyl-, sodium salt, unii-7cku5w4trm, p-toluenesulfinic acid, sodium salt, p-toluensulfinan sodny czech |
| PubChem CID | 2723791 |
| IUPAC Name | sodium;4-methylbenzenesulfinate |
| SMILES | CC1=CC=C(C=C1)S(=O)[O-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC434530250
|
N/A | 25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC434531000
|
N/A | 100g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 824-79-3 | |
| 178.18 | |
| sodium 4-methylbenzenesulfinate, sodium p-toluenesulfinate, p-toluenesulfinic acid sodium salt, sodium toluene-4-sulphinate, sodium p-tolylsulfinate, sodium 4-toluenesulfinate, benzenesulfinic acid, 4-methyl-, sodium salt, unii-7cku5w4trm, p-toluenesulfinic acid, sodium salt, p-toluensulfinan sodny czech | |
| sodium;4-methylbenzenesulfinate |
| C7H7O2S·Na | |
| KFZUDNZQQCWGKF-UHFFFAOYSA-M | |
| 2723791 | |
| CC1=CC=C(C=C1)S(=O)[O-].[Na+] |
Specifications
| 824-79-3 | |
| 96% min. (HPLC) | |
| sodium 4-methylbenzenesulfinate, sodium p-toluenesulfinate, p-toluenesulfinic acid sodium salt, sodium toluene-4-sulphinate, sodium p-tolylsulfinate, sodium 4-toluenesulfinate, benzenesulfinic acid, 4-methyl-, sodium salt, unii-7cku5w4trm, p-toluenesulfinic acid, sodium salt, p-toluensulfinan sodny czech | |
| KFZUDNZQQCWGKF-UHFFFAOYSA-M | |
| sodium;4-methylbenzenesulfinate | |
| 2723791 | |
| 97% |
| Authentic | |
| C7H7O2S·Na | |
| Solubility in water: soluble. | |
| CC1=CC=C(C=C1)S(=O)[O-].[Na+] | |
| 178.18 | |
| 178.18 | |
| p-Toluenesulfinic acid, sodium salt |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 212-538-5
RUO – Research Use Only