missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfinic acid, sodium salt, 97%, anhydrous

Catalog No. AC434530250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434530250 25 g Glass bottle
AC434531000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434530250 Supplier Thermo Scientific Chemicals Supplier No. 434530250

CAS: 824-79-3 | C7H7O2S·Na | 178.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 824-79-3
Molecular Formula C7H7O2S·Na
Molecular Weight (g/mol) 178.18
InChI Key KFZUDNZQQCWGKF-UHFFFAOYSA-M
Synonym sodium 4-methylbenzenesulfinate, sodium p-toluenesulfinate, p-toluenesulfinic acid sodium salt, sodium toluene-4-sulphinate, sodium p-tolylsulfinate, sodium 4-toluenesulfinate, benzenesulfinic acid, 4-methyl-, sodium salt, unii-7cku5w4trm, p-toluenesulfinic acid, sodium salt, p-toluensulfinan sodny czech
PubChem CID 2723791
IUPAC Name sodium;4-methylbenzenesulfinate
SMILES CC1=CC=C(C=C1)S(=O)[O-].[Na+]

Specifications

Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: soluble.
Formula Weight 178.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material p-Toluenesulfinic acid, sodium salt
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-538-5

RUO – Research Use Only