missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Tolualdehyde 95.0+%, TCI America™
Supplier: TCI America T107325ML
Specifications
| p-Tolualdehyde | |
| Yellow | |
| C8H8O | |
| 25 mL | |
| FXLOVSHXALFLKQ-UHFFFAOYSA-N | |
| 4-methylbenzaldehyde | |
| 7725 | |
| 120.15 | |
| Liquid |
| 104-87-0 | |
| 204°C | |
| MFCD00006954 | |
| p-tolualdehyde, 4-tolualdehyde, p-formyltoluene, p-tolylaldehyde, p-toluylaldehyde, p-methylbenzaldehyde, para-tolualdehyde, benzaldehyde, 4-methyl, para-methylbenzaldehyde, para-toluyl aldehyde | |
| CC1=CC=C(C=C1)C=O | |
| 120.151 | |
| CHEBI:28617 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 104-87-0 | |
| 120.151 | |
| FXLOVSHXALFLKQ-UHFFFAOYSA-N | |
| 7725 | |
| 4-methylbenzaldehyde |
| C8H8O | |
| MFCD00006954 | |
| p-tolualdehyde, 4-tolualdehyde, p-formyltoluene, p-tolylaldehyde, p-toluylaldehyde, p-methylbenzaldehyde, para-tolualdehyde, benzaldehyde, 4-methyl, para-methylbenzaldehyde, para-toluyl aldehyde | |
| CHEBI:28617 | |
| CC1=CC=C(C=C1)C=O |
Safety and Handling
EINECSNumber : (3)-2659
RTECSNumber : CU7034500
TSCA : Yes