missing translation for 'onlineSavingsMsg'
Learn More

p-Thiocresol, 98%

Catalog No. p-7027908
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2497014 25 g
AAB2497022 100 g
AAB2497036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2497014 Supplier Thermo Scientific Chemicals Supplier No. B2497014

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 106-45-6 | C7H8S | 124.201 g/mol

p-Thiocresol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Thiocresol is used as a pharmaceutical intermediate.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents, air.

Chemical Identifiers

CAS 106-45-6
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.201
MDL Number MFCD00004851
InChI Key WLHCBQAPPJAULW-UHFFFAOYSA-N
Synonym p-toluenethiol, p-thiocresol, 4-thiocresol, 4-methylthiophenol, p-methylthiophenol, benzenethiol, 4-methyl, 4-toluenethiol, p-tolyl mercaptan, p-thiolcresol, p-tolylthiol
PubChem CID 7811
IUPAC Name 4-methylbenzenethiol
SMILES CC1=CC=C(C=C1)S

Specifications

Melting Point 40°C to 44°C
Boiling Point 195°C
Flash Point 68°C (154°F)
Odor Stench
Quantity 25 g
UN Number UN2811
Beilstein 605761
Sensitivity Air Sensitive
Merck Index 14,9325
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 124.21
Percent Purity 98%
Chemical Name or Material p-Thiocresol
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 203-399-1
RTECSNumber XT8925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only