missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Thiocresol, 98%
Description
p-Thiocresol is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Thiocresol is used as a pharmaceutical intermediate.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Air sensitive. Store away from oxidizing agents, air.
p-Thiocresol is used as a pharmaceutical intermediate.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Air sensitive. Store away from oxidizing agents, air.
Specifications
Specifications
| Melting Point | 40°C to 44°C |
| Boiling Point | 195°C |
| Flash Point | 68°C (154°F) |
| Odor | Stench |
| Quantity | 100 g |
| UN Number | UN2811 |
| Beilstein | 605761 |
| Sensitivity | Air Sensitive |
| Merck Index | 14,9325 |
| Solubility Information | Soluble in alcohol. Insoluble in water. |
| Show More |
Safety and Handling
| Hazard Category | H302+H312+H332-H315-H319-H335 |
| Hazard Statement | GHS H Statement H331-H302-H312-H315-H319-H335 Toxic if inhaled. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 203-399-1 |
| RTECSNumber | XT8925000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only