missing translation for 'onlineSavingsMsg'
Learn More

p-Terphenyl 99.0+%, TCI America™

Supplier:  TCI America T0020100G

 View more versions of this product

Catalog No. T0020100G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

389°C
211°C
92-94-4
100 g
MFCD00003061
XJKSTNDFUHDPQJ-UHFFFAOYSA-N
1,4-diphenylbenzene
7115
230.31
p-Terphenyl
Crystal-Powder at 20°C
White/Yellow
C18H14
1,4-Diphenylbenzene
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3
230.31
CHEBI:52242
≥99.0% (GC)

Chemical Identifiers

92-94-4
230.31
XJKSTNDFUHDPQJ-UHFFFAOYSA-N
7115
1,4-diphenylbenzene
C18H14
MFCD00003061
1,4-Diphenylbenzene
CHEBI:52242
C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0017

RTECSNumber : WZ6475000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions