missing translation for 'onlineSavingsMsg'
Learn More

p-Terphenyl 99.0+%, TCI America™

Catalog No. p-7136035
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T002025G 25 g
T0020100G 100 g
T0020500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T002025G Supplier TCI America Supplier No. T002025G
Only null left

Chemical Identifiers

CAS 92-94-4
Molecular Formula C18H14
Molecular Weight (g/mol) 230.31
MDL Number MFCD00003061
InChI Key XJKSTNDFUHDPQJ-UHFFFAOYSA-N
Synonym 1,4-Diphenylbenzene
PubChem CID 7115
ChEBI CHEBI:52242
IUPAC Name 1,4-diphenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3

Specifications

Boiling Point 389°C
Melting Point 211°C
Physical Form Crystal-Powder at 20°C
Color White/Yellow
Quantity 25 g
Formula Weight 230.31
Percent Purity ≥99.0% (GC)
Chemical Name or Material p-Terphenyl
EINECSNumber (4)-0017
RTECSNumber WZ6475000
TSCA Yes