missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals p-Terphenyl, 99+%, pure

Catalog No. AC137920250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC137920250 25 g Plastic bottle
AC137921000 100 g Plastic bottle
AC137925000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC137920250 Supplier Thermo Scientific Chemicals Supplier No. 137920250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 92-94-4 | C18H14 | 230.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 92-94-4
Molecular Formula C18H14
Molecular Weight (g/mol) 230.31
MDL Number MFCD00003061
InChI Key XJKSTNDFUHDPQJ-UHFFFAOYSA-N
Synonym p-terphenyl, 1,1':4',1-terphenyl, p-diphenylbenzene, p-triphenyl, santowax p, 4-phenylbiphenyl, 4-phenyldiphenyl, para-terphenyl, biphenyl, 4-phenyl, pyrogallol tannin
PubChem CID 7115
ChEBI CHEBI:52242
IUPAC Name 1,4-diphenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3

Specifications

Melting Point 209.0°C to 216.0°C
Color White to Beige
Density 207g/mL
Boiling Point 383.0°C
Flash Point 207°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5C6H4C6H5
Quantity 25 g
Beilstein 05, 695
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,benzene and toluene
Formula Weight 230.31
Percent Purity 99+%
Grade Pure
Physical Form Crystalline Powder, Shiny Crystals or Flakes
Chemical Name or Material p-Terphenyl, Pure
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
EINECSNumber 202-205-2

RUO – Research Use Only