missing translation for 'onlineSavingsMsg'
Learn More

p-Sexiphenyl, TCI America™

Catalog No. p-7135839
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0220100MG 100 mg
S02201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0220100MG Supplier TCI America Supplier No. S0220100MG
Only null left

Chemical Identifiers

CAS 4499-83-6
Molecular Formula C36H26
Molecular Weight (g/mol) 458.604
MDL Number MFCD00039559
InChI Key ZEMDSNVUUOCIED-UHFFFAOYSA-N
Synonym p-sexiphenyl, p-hexaphenyl, p-quaterphenyl, 4,4'-diphenyl, 1-phenyl-4-4-4-4-phenylphenyl phenyl phenyl benzene, p-sexiphenylene, acmc-209k0d, phthalocyanine lead ii purified by sublimation, 1,1':4',1:4,1'-quaterphenyl, 4,4'-diphenyl, 4,4'-bis 1,1'-biphenyl-4-yl-1,1'-biphenyl
PubChem CID 78254
IUPAC Name 1-phenyl-4-[4-[4-(4-phenylphenyl)phenyl]phenyl]benzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)C4=CC=C(C=C4)C5=CC=C(C=C5)C6=CC=CC=C6

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 458.60
Physical Form Powder or Flakes
Chemical Name or Material p-Sexiphenyl