missing translation for 'onlineSavingsMsg'
Learn More

p-Quinquephenyl, TCI America™

Catalog No. p-7135622
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q0018100MG 100 mg
Q00181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q0018100MG Supplier TCI America Supplier No. Q0018100MG
Only null left

Chemical Identifiers

CAS 3073-05-0
Molecular Formula C30H22
Molecular Weight (g/mol) 382.506
MDL Number MFCD00059012
InChI Key OMCUOJTVNIHQTI-UHFFFAOYSA-N
Synonym p-quinquephenyl, p-pentaphenyl, 1,1':4',1:4,1':4',1-quinquephenyl, p-quinquiphenyl, acmc-209hie, 1,4-bis 4-phenylphenyl benzene, 1-phenyl-4-4-4-phenylphenyl phenyl benzene, 1,1:4,1':4',1-quinquephenyl, 4-1,1'-biphenyl-4-yl-4'-phenyl-1,1'-biphenyl
PubChem CID 137813
IUPAC Name 1,4-bis(4-phenylphenyl)benzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)C4=CC=C(C=C4)C5=CC=CC=C5

Specifications

Color White
Quantity 100 mg
Formula Weight 382.51
Physical Form Crystalline Powder
Chemical Name or Material p-Quinquephenyl