missing translation for 'onlineSavingsMsg'
Learn More

p-Quaterphenyl 98.0+%, TCI America™

Catalog No. Q00011G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00011G 1 g
Q00015G 5 g
Q000125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. Q00011G Supplier TCI America Supplier No. Q00011G

Chemical Identifiers

CAS 135-70-6
Molecular Formula C24H18
Molecular Weight (g/mol) 306.408
MDL Number MFCD00003062
InChI Key GPRIERYVMZVKTC-UHFFFAOYSA-N
Synonym p-quaterphenyl, 1,1':4',1:4,1'-quaterphenyl, benzerythrene, quadriphenyl, p-tetraphenyl, 4,4'-diphenylbiphenyl, p,p'-quaterphenyl, 4-quaterphenyl, unii-g8aqm6d0rk, 1-phenyl-4-4-phenylphenyl benzene
PubChem CID 8677
ChEBI CHEBI:52240
IUPAC Name 1-phenyl-4-(4-phenylphenyl)benzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)C4=CC=CC=C4

Specifications

Color White
Quantity 1 g
Formula Weight 306.41
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material p-Quaterphenyl