missing translation for 'onlineSavingsMsg'
Learn More

p-Phenylene diisothiocyanate, 99%

Catalog No. p-183028
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC417510050 5 g Glass bottle
AC417510250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC417510050 Supplier Thermo Scientific Chemicals Supplier No. 417510050
Only null left

CAS: 4044-65-9 | C8H4N2S2 | 192.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4044-65-9
Molecular Formula C8H4N2S2
Molecular Weight (g/mol) 192.27
MDL Number MFCD00004811
InChI Key OMWQUXGVXQELIX-UHFFFAOYSA-N
Synonym bitoscanate, 1,4-phenylene diisothiocyanate, jonit, p-phenylene diisothiocyanate, benzene, 1,4-diisothiocyanato, bitoscanat, bitoscanate inn, phenylene thiocyanate, bitoscanatum latin, 1,4-phenylenediisothiocyanate
PubChem CID 19958
IUPAC Name 1,4-diisothiocyanatobenzene
SMILES C1=CC(=CC=C1N=C=S)N=C=S

Specifications

Melting Point 129.0°C to 133.0°C
Color Beige-Gray to White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula SCNC6H4NCS
Quantity 5 g
Merck Index 15, 1309
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and chloroform,slightly soluble in acetone
Formula Weight 192.27
Percent Purity 99%
Physical Form Crystalline Needles or Powder
Chemical Name or Material p-Phenylene diisothiocyanate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep a
EINECSNumber 223-741-3
RTECSNumber NX9150000
TSCA TSCA

RUO – Research Use Only