missing translation for 'onlineSavingsMsg'
Learn More

p-Phenetidine 98.0+%, TCI America™

Catalog No. P009025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P009025G 25 g
P0090500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P009025G Supplier TCI America Supplier No. P009025G
Only null left

p-Phenetidine 98.0+%

Chemical Identifiers

CAS 156-43-4
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00007865
InChI Key IMPPGHMHELILKG-UHFFFAOYSA-N
Synonym p-phenetidine, phenetidine, 4-aminophenetole, p-ethoxyaniline, p-phenetidin, benzenamine, 4-ethoxy, phenethidine, p-aminophenetole, 4-ethoxybenzenamine, 1-amino-4-ethoxybenzene
PubChem CID 9076
ChEBI CHEBI:85505
IUPAC Name 4-ethoxyaniline
SMILES CCOC1=CC=C(N)C=C1

Specifications

Melting Point 5°C
Color Yellow
Boiling Point 254°C
Quantity 25 g
UN Number 2311
Formula Weight 137.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material p-Phenetidine
EINECSNumber (3)-0682
RTECSNumber SI6465500
TSCA Yes
Recommended Storage Refrigerator