missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Nitro Blue Tetrazolium Chloride, Molecular Biology Reagent Grade, MP Biomedicals™
$167.68 - $167.68
Chemical Identifiers
| CAS | 298-83-9 |
|---|---|
| Molecular Formula | C40H30Cl2N10O6 |
| Molecular Weight (g/mol) | 817.644 |
| MDL Number | MFCD00012159 |
| InChI Key | FSVCQIDHPKZJSO-UHFFFAOYSA-L |
| Synonym | nitrotetrazolium blue chloride, nitro bt, nitro blue tetrazolium, nbt, nitro blue tetrazolium chloride, p-nitrotetrazolium blue, nitrotetrazolium blue, nitroblue tetrazolium chloride, p-nitro blue tetrazolium chloride, nitro tetrazolium bt |
| PubChem CID | 9281 |
| ChEBI | CHEBI:9505 |
| IUPAC Name | 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride |
| SMILES | COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19399983
|
MP Biomedicals Inc
0219399983 |
250 mg |
Each for $167.68
|
|
|||||
Chemical Identifiers
| 298-83-9 | |
| 817.644 | |
| FSVCQIDHPKZJSO-UHFFFAOYSA-L | |
| 9281 | |
| 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride |
| C40H30Cl2N10O6 | |
| MFCD00012159 | |
| nitrotetrazolium blue chloride, nitro bt, nitro blue tetrazolium, nbt, nitro blue tetrazolium chloride, p-nitrotetrazolium blue, nitrotetrazolium blue, nitroblue tetrazolium chloride, p-nitro blue tetrazolium chloride, nitro tetrazolium bt | |
| CHEBI:9505 | |
| COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-] |
Specifications
| 298-83-9 | |
| 0.981 | |
| MFCD00012159 | |
| nitrotetrazolium blue chloride, nitro bt, nitro blue tetrazolium, nbt, nitro blue tetrazolium chloride, p-nitrotetrazolium blue, nitrotetrazolium blue, nitroblue tetrazolium chloride, p-nitro blue tetrazolium chloride, nitro tetrazolium bt | |
| COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-] | |
| 817.644 | |
| CHEBI:9505 | |
| Molecular Biology Reagent | |
| p-Nitro Blue Tetrazolium Chloride, For HPLC analysis |
| Yellow | |
| C40H30Cl2N10O6 | |
| 250 mg | |
| FSVCQIDHPKZJSO-UHFFFAOYSA-L | |
| 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride | |
| 9281 | |
| 817.6 | |
| Powder |
Safety and Handling
EINECSNumber : 206-067-4
Recommended Storage : Store at +4°C.