Learn More
p-Methylstyrene, 97%, stabilized, ACROS Organics™
Supplier: Thermo Scientific Chemicals 127700025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| p-Methylstyrene | |
| 0.8960g/mL | |
| 45°C | |
| 97% min. (GC) | |
| C9H10 | |
| CH3C6H4CH=CH2 | |
| 2.5L | |
| 0.896 | |
| Solubility in water: insoluble | |
| CC1=CC=C(C=C)C=C1 | |
| 118.18 | |
| 12161 | |
| 97% |
| 622-97-9 | |
| 170.0°C to 175.0°C | |
| Authentic | |
| Glass bottle | |
| 1.5410 to 1.5430 | |
| MFCD00008621 | |
| 05,485 | |
| 4-methylstyrene, 1-methyl-4-vinylbenzene, p-methylstyrene, 4-vinyltoluene, p-vinyltoluene, benzene, 1-ethenyl-4-methyl, styrene, p-methyl, p-methyl styrene, 1-p-tolylethene, para-methylstyrene | |
| JLBJTVDPSNHSKJ-UHFFFAOYSA-N | |
| 1-ethenyl-4-methylbenzene | |
| 0.1% 3, 5-di-tert.-butylcatechol | |
| 118.18 |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause genetic defects.
Toxic to aquatic life with long lasting effects.
Ma
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o
GHS Signal Word: Danger
EINECSNumber : 210-762-8
RUO â Research Use Only