Learn More
p-Dithiane-2,5-diol, 96%
CAS: 40018-26-6 | C4H8O2S2 | 152.23 g/mol
$320.04 - $320.04
Chemical Identifiers
| CAS | 40018-26-6 |
|---|---|
| Molecular Formula | C4H8O2S2 |
| Molecular Weight (g/mol) | 152.23 |
| InChI Key | YUIOPHXTILULQC-UHFFFAOYSA-N |
| Synonym | p-dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithiane, mercaptoacetaldehyde dimer, 4-dithiane-2,5-diol, 1,4 dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithian, p-dthane-2,5-dol, pubchem20480, 1,4dithiane-2,5diol, acmc-1anme |
| PubChem CID | 98330 |
| IUPAC Name | 1,4-dithiane-2,5-diol |
| SMILES | C1C(SCC(S1)O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC179932500
|
Thermo Scientific Chemicals
179932500 |
250 g | Plastic bottle |
Each for $320.04
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 40018-26-6 | |
| 152.23 | |
| p-dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithiane, mercaptoacetaldehyde dimer, 4-dithiane-2,5-diol, 1,4 dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithian, p-dthane-2,5-dol, pubchem20480, 1,4dithiane-2,5diol, acmc-1anme | |
| 1,4-dithiane-2,5-diol |
| C4H8O2S2 | |
| YUIOPHXTILULQC-UHFFFAOYSA-N | |
| 98330 | |
| C1C(SCC(S1)O)O |
Specifications
| 40018-26-6 | |
| White to Yellow | |
| 95% min. (GC) | |
| C4H8O2S2 | |
| p-dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithiane, mercaptoacetaldehyde dimer, 4-dithiane-2,5-diol, 1,4 dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithian, p-dthane-2,5-dol, pubchem20480, 1,4dithiane-2,5diol, acmc-1anme | |
| YUIOPHXTILULQC-UHFFFAOYSA-N | |
| 1,4-dithiane-2,5-diol | |
| 98330 | |
| 96% | |
| p-Dithiane-2,5-diol |
| 130.0°C | |
| Authentic | |
| Plastic bottle | |
| 250 g | |
| Solubility in water: 3.9g/L (20°C). | |
| C1C(SCC(S1)O)O | |
| 152.23 | |
| 152.23 | |
| Crystalline Powder |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Causes serious eye damage.
Toxic if swallowed.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
GHS Signal Word: Danger
EINECSNumber : 254-751-6
RUO – Research Use Only