accessibility menu, dialog, popup

UserName

CAS RN 40018-26-6

IUPAC Name: 1,4-dithiane-2,5-diol
Synonyms: 1,4-dithiane-2,5-diol
Molecular Weight (g/mol): 152.23
Molecular Formula: C4H8O2S2
InChi Key: YUIOPHXTILULQC-UHFFFAOYNA-N
SMILES: OC1CSC(O)CS1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)