missing translation for 'onlineSavingsMsg'
Learn More

p-Dimethylaminobenzaldehyde (Certified ACS), Fisher Chemical™

Catalog No. p-114912
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
Catalog No. Quantity Packaging
D71100 100 g Glass Bottle
1 options

Catalog No. D71100

Supplier: Thermo Fisher Scientific D71 100

Only null left

Chemical Identifiers

CAS 100-10-7
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00003381
InChI Key BGNGWHSBYQYVRX-UHFFFAOYSA-N
Synonym 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
PubChem CID 7479
IUPAC Name 4-(dimethylamino)benzaldehyde
SMILES CN(C)C1=CC=C(C=O)C=C1

Specifications

Alpha Vector Dimethylaminobenzaldehyde
Melting Point 70°C
Color Yellow
Density 5.15 (Air=1.0)
Boiling Point 176°C
Flash Point 164°C
Ignition Residue 0.1% max.
Packaging Glass Bottle
Linear Formula (CH3)2NC6H4CHO
Quantity 100 g
Solubility Information Pass Test (in HCl)
Formula Weight 149.19
Grade Certified ACS
Physical Form Powder or Solid
Chemical Name or Material p-Dimethylaminobenzaldehyde
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Harmful if swallowed. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:1
Instability:1