missing translation for 'onlineSavingsMsg'
Learn More

p-(Dimethylamino)benzaldehyde, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02003566
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003566 25 g Poly Bottle
02003567 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02003566 Supplier Avantor Supplier No. J41803

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 100-10-7
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00003381
InChI Key BGNGWHSBYQYVRX-UHFFFAOYSA-N
Synonym 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
PubChem CID 7479
IUPAC Name 4-(dimethylamino)benzaldehyde
SMILES CN(C)C1=CC=C(C=O)C=C1

Specifications

Packaging Poly Bottle
Quantity 25 g
Formula Weight 149.19
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material p-(Dimethylamino)benzaldehyde