missing translation for 'onlineSavingsMsg'
Learn More

p-Dimethylamino)benzaldehyde ACS AR, Macron Fine Chemicals™

macron

$409.88 - $409.88

Chemical Identifiers

CAS 100-10-7
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00003381
InChI Key BGNGWHSBYQYVRX-UHFFFAOYSA-N
Synonym 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
PubChem CID 7479
IUPAC Name 4-(dimethylamino)benzaldehyde
SMILES CN(C)C1=CC=C(C=O)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
MK1836100
View Documents
Avantor Macron
183657
100 g Glass
Each for $409.88
Only null left
 
Description

Description

Specifications

Chemical Identifiers

100-10-7
149.19
BGNGWHSBYQYVRX-UHFFFAOYSA-N
7479
CN(C)C1=CC=C(C=O)C=C1
C9H11NO
MFCD00003381
4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
4-(dimethylamino)benzaldehyde

Specifications

100-10-7
C9H11NO
100 g
BGNGWHSBYQYVRX-UHFFFAOYSA-N
4-(dimethylamino)benzaldehyde
7479
ACS
Glass
MFCD00003381
4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
CN(C)C1=CC=C(C=O)C=C1
149.19
149.19
p-(Dimethylamino)benzaldehyde
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'