missing translation for 'onlineSavingsMsg'
Learn More

p-Cymene, 99+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 111760100

Catalog No. AC111760100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

p-Cymene
0.8540g/mL
47°C
99% min. (GC)
C10H14
CH3C6H4CH(CH3)2
10L
0.854
p-cymene, 4-isopropyltoluene, p-isopropyltoluene, dolcymene, p-cymol, paracymene, cymene, para-cymene, 1-isopropyl-4-methylbenzene, camphogen
HFPZCAJZSCWRBC-UHFFFAOYSA-N
1-methyl-4-propan-2-ylbenzene
7463
134.22
99-87-6
176.0°C to 178.0°C
Authentic
Plastic drum
1.4890 to 1.4910
MFCD00008893
05, II, 322
15, 2760
Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,ether,chloroform,acetone,,benzene and ccl4
CC1=CC=C(C=C1)C(C)C
134.22
CHEBI:28768
99+%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
If skin irritation occurs: Get medical advice/attentio

GHS Signal Word: Danger

EINECSNumber : 202-796-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only