missing translation for 'onlineSavingsMsg'
Learn More

p-Cymene, 97%

CAS: 99-87-6 | C10H14 | 134.222 g/mol

$60.41 - $175.84

Chemical Identifiers

CAS 99-87-6
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00008893
InChI Key HFPZCAJZSCWRBC-UHFFFAOYSA-N
Synonym p-cymene, 4-isopropyltoluene, p-isopropyltoluene, dolcymene, p-cymol, paracymene, cymene, para-cymene, 1-isopropyl-4-methylbenzene, camphogen
PubChem CID 7463
ChEBI CHEBI:28768
IUPAC Name 1-methyl-4-propan-2-ylbenzene
SMILES CC1=CC=C(C=C1)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA19226AP
View Documents Promotion Details
N/A 500 mL
Each for $60.41
Only null left
 
AAA192260F
View Documents Promotion Details
N/A 2500 mL
Each for $175.84
Only null left
 
Description

Description

It is used in heat-exchange media, metal polishes, and thinners for lacquers/varnishes. Also used to make para-cresol, carvacrol, and synthetic resins. It is also used as a component of commercial terpene solvent mixtures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in heat-exchange media, metal polishes, and thinners for lacquers/varnishes. Also used to make para-cresol, carvacrol, and synthetic resins. It is also used as a component of commercial terpene solvent mixtures.

Solubility
Soluble in water at 0.023g/L.20°C.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

99-87-6
134.222
HFPZCAJZSCWRBC-UHFFFAOYSA-N
7463
1-methyl-4-propan-2-ylbenzene
C10H14
MFCD00008893
p-cymene, 4-isopropyltoluene, p-isopropyltoluene, dolcymene, p-cymol, paracymene, cymene, para-cymene, 1-isopropyl-4-methylbenzene, camphogen
CHEBI:28768
CC1=CC=C(C=C1)C(C)C

Specifications

99-87-6
0.86
47°C (116°F)
C10H14
MFCD00008893
1903377
p-cymene, 4-isopropyltoluene, p-isopropyltoluene, dolcymene, p-cymol, paracymene, cymene, para-cymene, 1-isopropyl-4-methylbenzene, camphogen
HFPZCAJZSCWRBC-UHFFFAOYSA-N
1-methyl-4-propan-2-ylbenzene
7463
134.22
p-Cymene
-68°C
176°C to 178°C
Aromatic
1.49
UN2046
14,2763
Soluble in water at 0.023g/L.20°C.
CC1=CC=C(C=C1)C(C)C
134.222
CHEBI:28768
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c

H226-H304-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CYMENES

EINECSNumber : 202-796-7

RTECSNumber : GZ5950000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only