missing translation for 'onlineSavingsMsg'
Learn More

p-Benzoquinone dioxime, 95%

Catalog No. aaa175320b
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA175320B 1000 g
AAA1753218 50 g
AAA1753230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA175320B Supplier Thermo Scientific Chemicals Supplier No. A175320B
Only null left

CAS: 105-11-3 | C6H6N2O2 | 138.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105-11-3
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00063636
InChI Key LNHURPJLTHSVMU-CGXWXWIYSA-N
Synonym 1,4-benzoquinone dioxime, p-benzoquinonedioxime, p-benzoquinone dioxime, p-quinone dioxime, quinone dioxime, benzoquinone dioxime, p-quinone oxime, actor q, dioxime p-benzoquinone, dibenzo pqd
PubChem CID 7737
IUPAC Name N-(4-nitrosophenyl)hydroxylamine
SMILES ON=C1C=CC(C=C1)=NO

Specifications

Melting Point ∼223°C (decomposition)
Density 1.49
Quantity 1000 g
Beilstein 2043234
Formula Weight 138.13
Percent Purity 95%
Chemical Name or Material p-Benzoquinone dioxime
Hazard Category H302-H351
Hazard Statement GHS H Statement
H351-H302
Suspected of causing cancer.
Harmful if swallowed.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
EINECSNumber 203-271-5
RTECSNumber DK4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only