missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Anisil 98.0+%, TCI America™
$89.48 - $287.06
Chemical Identifiers
| CAS | 1226-42-2 |
|---|---|
| Molecular Formula | C16H14O4 |
| Molecular Weight (g/mol) | 270.284 |
| MDL Number | MFCD00008405 |
| InChI Key | YNANGXWUZWWFKX-UHFFFAOYSA-N |
| Synonym | 4,4'-dimethoxybenzil, anisil, p-anisil, 1,2-bis 4-methoxyphenyl ethane-1,2-dione, bis 4-methoxyphenyl ethanedione, ethanedione, bis 4-methoxyphenyl, di-p-anisoyl, p,p'-dimethoxybenzil, 1,2-ethanedione, 1,2-bis 4-methoxyphenyl, 1,2-bis 4-methoxyphenyl-1,2-ethanedione |
| PubChem CID | 71043 |
| IUPAC Name | 1,2-bis(4-methoxyphenyl)ethane-1,2-dione |
| SMILES | COC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)OC |
Chemical Identifiers
| 1226-42-2 | |
| 270.284 | |
| YNANGXWUZWWFKX-UHFFFAOYSA-N | |
| 71043 | |
| COC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)OC |
| C16H14O4 | |
| MFCD00008405 | |
| 4,4'-dimethoxybenzil, anisil, p-anisil, 1,2-bis 4-methoxyphenyl ethane-1,2-dione, bis 4-methoxyphenyl ethanedione, ethanedione, bis 4-methoxyphenyl, di-p-anisoyl, p,p'-dimethoxybenzil, 1,2-ethanedione, 1,2-bis 4-methoxyphenyl, 1,2-bis 4-methoxyphenyl-1,2-ethanedione | |
| 1,2-bis(4-methoxyphenyl)ethane-1,2-dione |
Specifications
| 1226-42-2 | |
| C16H14O4 | |
| 5 g | |
| YNANGXWUZWWFKX-UHFFFAOYSA-N | |
| 1,2-bis(4-methoxyphenyl)ethane-1,2-dione | |
| 71043 | |
| ≥98.0% (GC) | |
| p-Anisil |
| 133°C | |
| MFCD00008405 | |
| 4,4'-dimethoxybenzil, anisil, p-anisil, 1,2-bis 4-methoxyphenyl ethane-1,2-dione, bis 4-methoxyphenyl ethanedione, ethanedione, bis 4-methoxyphenyl, di-p-anisoyl, p,p'-dimethoxybenzil, 1,2-ethanedione, 1,2-bis 4-methoxyphenyl, 1,2-bis 4-methoxyphenyl-1,2-ethanedione | |
| COC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)OC | |
| 270.284 | |
| 270.28 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0277
TSCA : Yes