missing translation for 'onlineSavingsMsg'
Learn More

p-Anisidine 98.0+%, TCI America™

Catalog No. p-7116929
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A048725G 25 g
A0487500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A048725G Supplier TCI America Supplier No. A048725G
Only null left

Chemical Identifiers

CAS 104-94-9
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00007864
InChI Key BHAAPTBBJKJZER-UHFFFAOYSA-N
Synonym p-anisidine, 4-anisidine, 4-methoxybenzenamine, 4-aminoanisole, p-aminoanisole, para-anisidine, p-methoxyaniline, benzenamine, 4-methoxy, 4-methoxybenzeneamine, p-anisylamine
PubChem CID 7732
ChEBI CHEBI:82388
IUPAC Name 4-methoxyaniline
SMILES COC1=CC=C(C=C1)N

Specifications

Melting Point 59°C
Color White-Yellow
Boiling Point 246°C
Quantity 25 g
UN Number 2811
Formula Weight 123.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material p-Anisidine
EINECSNumber (3)-0682
RTECSNumber BZ5450000
TSCA Yes