missing translation for 'onlineSavingsMsg'
Learn More

p-Anisidine, 99%

Catalog No. p-61020
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104830050 5 g Glass bottle
AC104832500 250 g Glass bottle
AC104830010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC104830050 Supplier Thermo Scientific Chemicals Supplier No. 104830050
Only null left

CAS: 104-94-9 | C7H9NO | 123.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104-94-9 , 90-04-0
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.15
MDL Number MFCD00007864
InChI Key BHAAPTBBJKJZER-UHFFFAOYSA-N
Synonym p-anisidine, 4-anisidine, 4-methoxybenzenamine, 4-aminoanisole, p-aminoanisole, para-anisidine, p-methoxyaniline, benzenamine, 4-methoxy, 4-methoxybenzeneamine, p-anisylamine
PubChem CID 7732
ChEBI CHEBI:82388
IUPAC Name 4-methoxyaniline
SMILES COC1=CC=C(C=C1)N

Specifications

Melting Point 56.0°C to 59.0°C
Color Brown to Gray
Density 1.0600g/mL
Boiling Point 240.0°C to 243.0°C
Flash Point 122°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4NH2
Quantity 5 g
Beilstein 13, 435
Specific Gravity 1.06
Merck Index 14, 667
Solubility Information Solubility in water: 21g/L (20°C). Other solubilities: freely soluble in methanol, ethanol and ether, soluble in acetone and benzene
Formula Weight 123.15
Percent Purity 99%
Physical Form Crystalline Powder, Granules or Flakes
Chemical Name or Material p-Anisidine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
May cause cancer.
Fatal if inhaled.
Fatal in contact with skin.
Fatal if swallowed.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Rinse mouth.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 203-254-2
RTECSNumber BZ5450000
TSCA TSCA

RUO – Research Use Only