missing translation for 'onlineSavingsMsg'
Learn More

p-Anisidine, 99%

CAS: 104-94-9 | C7H9NO | 123.155 g/mol

Supplier:  Thermo Scientific Chemicals A109460B

 View more versions of this product

Catalog No. AAA109460B


Only null left
Explore available promotions

Description

Description

p-Anisidine is used in the production of azo dyes. It is also used for biochemical research and in organic synthesis. Further, it is used for detecting unsaturated aldehydes. It is also employed in food quality testing. In addition to this, it is used in the preparation of 1-methoxy-4-trans-styryl-benzene by reacting with vinylbenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Anisidine is used in the production of azo dyes. It is also used for biochemical research and in organic synthesis. Further, it is used for detecting unsaturated aldehydes. It is also employed in food quality testing. In addition to this, it is used in the preparation of 1-methoxy-4-trans-styryl-benzene by reacting with vinylbenzene.

Solubility
Soluble in acetone, benzene, ether and ethanol. Insoluble in water.

Notes
Air and light sensitive. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides and chloroformates.
Specifications

Specifications

p-Anisidine
56°C to 60°C
240°C to 243°C
C7H9NO
1000 g
471556
p-anisidine, 4-anisidine, 4-methoxybenzenamine, 4-aminoanisole, p-aminoanisole, para-anisidine, p-methoxyaniline, benzenamine, 4-methoxy, 4-methoxybenzeneamine, p-anisylamine
BHAAPTBBJKJZER-UHFFFAOYSA-N
4-methoxyaniline
7732
123.16
104-94-9
1.089
122°C (251°F)
MFCD00007864
UN2811
14,667
Soluble in acetone,benzene,ether and ethanol. Insoluble in water.
COC1=CC=C(C=C1)N
123.155
CHEBI:82388
99%

Chemical Identifiers

104-94-9
123.155
BHAAPTBBJKJZER-UHFFFAOYSA-N
7732
4-methoxyaniline
C7H9NO
MFCD00007864
p-anisidine, 4-anisidine, 4-methoxybenzenamine, 4-aminoanisole, p-aminoanisole, para-anisidine, p-methoxyaniline, benzenamine, 4-methoxy, 4-methoxybenzeneamine, p-anisylamine
CHEBI:82388
COC1=CC=C(C=C1)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P201-P202-P260-P262-P264b-P270-P271-P281-P284-P301+P310-P302+P350-P304+P340-P308+P313-P310-P330-P361-P363-P501c

H300+H310+H330-H350-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 203-254-2

RTECSNumber : BZ5450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only