missing translation for 'onlineSavingsMsg'
Learn More

p-Anisic Acid 99.0+%, TCI America™

Catalog No. p-7116925
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
A048225G 25 g
A0482500G 500 g
2 options

Catalog No. A048225G

Supplier: TCI America A048225G

Only null left

Chemical Identifiers

CAS 100-09-4
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002542
InChI Key ZEYHEAKUIGZSGI-UHFFFAOYSA-N
Synonym p-anisic acid, anisic acid, p-methoxybenzoic acid, draconic acid, 4-anisic acid, benzoic acid, 4-methoxy, anisic acid, para, 4-methoxybenzoate, para-anisic acid, anisic acid, p-isomer
PubChem CID 7478
ChEBI CHEBI:40813
IUPAC Name 4-methoxybenzoic acid
SMILES COC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 184°C
Color White
Boiling Point 275°C
Quantity 25 g
Formula Weight 152.15
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material p-Anisic Acid
EINECSNumber (3)-1659
RTECSNumber BZ4395000
TSCA Yes