missing translation for 'onlineSavingsMsg'
Learn More

p-Anisic acid, 98%

Catalog No. p-61006
Change view
Click to view available options
Quantity:
100 g
250 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC152371000 100 g Plastic bottle
AC152372500 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC152371000 Supplier Thermo Scientific Chemicals Supplier No. 152371000

CAS: 100-09-4 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100-09-4
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002542
InChI Key ZEYHEAKUIGZSGI-UHFFFAOYSA-N
Synonym p-anisic acid, anisic acid, p-methoxybenzoic acid, draconic acid, 4-anisic acid, benzoic acid, 4-methoxy, anisic acid, para, 4-methoxybenzoate, para-anisic acid, anisic acid, p-isomer
PubChem CID 7478
ChEBI CHEBI:40813
IUPAC Name 4-methoxybenzoic acid
SMILES COC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 181.0°C to 186.0°C
Color Beige-Gray to White
Boiling Point 275.0°C
Flash Point 185°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Refractive Index 1.5710 to 1.5760
Linear Formula CH3OC6H4CO2H
Quantity 100 g
Beilstein 10,IV,346
Merck Index 15,659
Solubility Information Solubility in water: 0.3g/L (20 c). Other solubilities: freely soluble in alcohol,ether,chloroform and,ethyl acetate
Formula Weight 152.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material p-Anisic acid, 98%
Show More Show Less
EINECSNumber 202-818-5
RTECSNumber BZ4395000
TSCA TSCA

RUO – Research Use Only