missing translation for 'onlineSavingsMsg'
Learn More

p-Anisaldehyde dimethyl acetal, 98%, Thermo Scientific™

Chemical Identifiers

CAS 2186-92-7
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.22
MDL Number MFCD00036507
InChI Key NNHYAHOTXLASEA-UHFFFAOYSA-N
Synonym 1-dimethoxymethyl-4-methoxybenzene, p-anisaldehyde dimethyl acetal, anisaldehyde dimethyl acetal, 4-methoxybenzaldehyde dimethyl acetal, p-dimethoxymethyl anisole, benzene, 1-dimethoxymethyl-4-methoxy, p-anisaldehydedimethylacetal, unii-xte77a5et4, anisicaldehyde dimethylacetal, p-anisaldehyde dimethylacetal
PubChem CID 75140
IUPAC Name 1-(dimethoxymethyl)-4-methoxybenzene
SMILES COC(OC)C1=CC=C(OC)C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC352290250
SDS View Documents
Thermo Scientific Chemicals
352290250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC352291000
SDS View Documents
Thermo Scientific Chemicals
352291000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC352295000
SDS View Documents
Thermo Scientific Chemicals
352295000
500g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2186-92-7
182.22
NNHYAHOTXLASEA-UHFFFAOYSA-N
75140
COC(OC)C1=CC=C(OC)C=C1
C10H14O3
MFCD00036507
1-dimethoxymethyl-4-methoxybenzene, p-anisaldehyde dimethyl acetal, anisaldehyde dimethyl acetal, 4-methoxybenzaldehyde dimethyl acetal, p-dimethoxymethyl anisole, benzene, 1-dimethoxymethyl-4-methoxy, p-anisaldehydedimethylacetal, unii-xte77a5et4, anisicaldehyde dimethylacetal, p-anisaldehyde dimethylacetal
1-(dimethoxymethyl)-4-methoxybenzene

Specifications

2186-92-7
235.0°C
Authentic
Glass bottle
1.5045 to 1.5065
25g
1-dimethoxymethyl-4-methoxybenzene, p-anisaldehyde dimethyl acetal, anisaldehyde dimethyl acetal, 4-methoxybenzaldehyde dimethyl acetal, p-dimethoxymethyl anisole, benzene, 1-dimethoxymethyl-4-methoxy, p-anisaldehydedimethylacetal, unii-xte77a5et4, anisicaldehyde dimethylacetal, p-anisaldehyde dimethylacetal
NNHYAHOTXLASEA-UHFFFAOYSA-N
1-(dimethoxymethyl)-4-methoxybenzene
75140
98%
1.0680g/mL
>110°C
97.5% min. (GC)
C10H14O3
MFCD00036507
1.068
Solubility in water: insoluble. Other solubilities: soluble in alcohol,soluble in common organic solvents
COC(OC)C1=CC=C(OC)C=C1
182.22
182.22
p-Anisaldehyde dimethyl acetal
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 218-577-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only