missing translation for 'onlineSavingsMsg'
Learn More

p-Anisaldehyde, Analytical Standard, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012817
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012817 1 mL
111012818 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. 111012817 missing translation for 'mfr' Merck Emd Millipore missing translation for 'supplierNo' 970631MLF

p-anisaldehyde is an aromatic aldehyde commonly found in anise seed oil. It shows acaricidal activity and is primarily used, as a lead compound for the development of new agents for the selective control of house dust mites.

Specifications

CAS 123-11-5
Vapor Density 4.7 (vs. air)
Density 1.119 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3OC6H4CHO
MDL Number MFCD00003385
Melting Point -1°C
Boiling Point 248°C
Synonym 4-Methoxybenzaldehyde; Aubépine
Quantity 1 mL
Formula Weight 136.15
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less