missing translation for 'onlineSavingsMsg'
Learn More

p-Aminobenzoic Acid, 99+%, MP Biomedicals

Catalog No. p-2625512
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19461901 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19461901 Supplier MP Biomedicals Inc Supplier No. 0219461901
Only null left

Chemical Identifiers

CAS 150-13-0
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007894
InChI Key ALYNCZNDIQEVRV-UHFFFAOYSA-N
Synonym p-aminobenzoic acid, paba, para-aminobenzoic acid, vitamin bx, sunbrella, p-carboxyaniline, 4-carboxyaniline, hachemina, paraminol, benzoic acid, 4-amino
PubChem CID 978
ChEBI CHEBI:30753
IUPAC Name 4-aminobenzoic acid
SMILES NC1=CC=C(C=C1)C(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 137.1
Grade Cell Culture Reagent
Physical Form Crystalline Powder
Chemical Name or Material p-Aminobenzoic Acid, 99+%, Free Acid
Recommended Storage Room temperature