missing translation for 'onlineSavingsMsg'
Learn More

p-Acetotoluidide, 99%

Catalog No. AC400161000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC400161000 100 g Glass bottle
AC400165000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC400161000 Supplier Thermo Scientific Chemicals Supplier No. 400161000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 103-89-9 | C9H11NO | 149.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103-89-9
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00008677
InChI Key YICAMJWHIUMFDI-UHFFFAOYSA-N
Synonym 4'-methylacetanilide, p-acetotoluidide, n-4-methylphenyl acetamide, 4-methylacetanilide, n-acetyl-p-toluidine, p-acetotoluide, acetyl-p-toluidine, p-acetotoluidine, acetamide, n-4-methylphenyl, 4-acetotoluide
PubChem CID 7684
IUPAC Name N-(4-methylphenyl)acetamide
SMILES CC1=CC=C(C=C1)NC(=O)C

Specifications

Melting Point 148.0°C to 151.0°C
Color Beige to Brown
Density 1.2100g/mL
Boiling Point 307.0°C
Flash Point 168°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4NHCOCH3
Quantity 100 g
Specific Gravity 1.21
Merck Index 15, 72
Solubility Information Solubility in water: 1g/l(25°C). Other solubilities: soluble in glacial acetic acid,soluble in alcohol,ether,ethyl acetate
Formula Weight 149.19
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material p-Acetotoluidide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-155-4

RUO – Research Use Only