missing translation for 'onlineSavingsMsg'
Learn More

p-Acetoacetophenetidide 98.0+%, TCI America™

Catalog No. A203925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A203925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A203925G Supplier TCI America Supplier No. A203925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 122-82-7
Molecular Formula C12H15NO3
Molecular Weight (g/mol) 221.26
MDL Number MFCD00043937
InChI Key WWROGCAUSKGAMX-UHFFFAOYSA-N
Synonym N-Acetoacetyl-p-phenetidine, N-Acetoacetyl-4-ethoxyaniline, 4′C-Ethoxyacetoacetanilide
PubChem CID 61053
IUPAC Name N-(4-ethoxyphenyl)-3-oxobutanamide
SMILES CCOC1=CC=C(NC(=O)CC(C)=O)C=C1

Specifications

Melting Point 104°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 221.26
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material p-Acetoacetophenetidide
EINECSNumber (3)-0698
RTECSNumber AK5800000
TSCA Yes