accessibility menu, dialog, popup

UserName

CAS RN 122-82-7

IUPAC Name: N-(4-ethoxyphenyl)-3-oxobutanamide
Synonyms: N-(4-ethoxyphenyl)-3-oxobutanamide
Molecular Weight (g/mol): 221.26
Molecular Formula: C12H15NO3
InChi Key: WWROGCAUSKGAMX-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(NC(=O)CC(C)=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results