missing translation for 'onlineSavingsMsg'
Learn More

Oxytetracycline Hydrochloride 95.0+%, TCI America™

Catalog No. p-7134356
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04755G 5 g
O047525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04755G Supplier TCI America Supplier No. O04755G
Only null left

Chemical Identifiers

CAS 2058-46-0
Molecular Formula C22H25ClN2O9
Molecular Weight (g/mol) 496.897
MDL Number MFCD00135815
InChI Key UTAHTGPLLFLKGW-IFLJXUKPSA-N
PubChem CID 121235537
IUPAC Name (4S,4aR,5S,5aR,6S,12aS)-4-(dimethylamino)-3,5,6,10,11,12a-hexahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2C(C3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O)O.Cl

Specifications

Melting Point 204°C
Color Yellow
Quantity 5 g
UN Number 3077
Formula Weight 496.90
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Oxytetracycline Hydrochloride
EINECSNumber (9)-0271
RTECSNumber QI8225000
TSCA Yes