missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Oxytetracycline Dihydrate 95%

$124.11 - $888.40

Chemical Identifiers

CAS 6153-64-6
Molecular Formula C22H28N2O11
Molecular Weight (g/mol) 496.469
MDL Number MFCD00151234
InChI Key IMLJLCJZQLGHJS-JEKSYDDFSA-N
PubChem CID 54680691
IUPAC Name (4S,4aR,5S,5aR,6S,12aR)-4-(dimethylamino)-1,5,6,10,11,12a-hexahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;dihydrate
SMILES CC1(C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O)O.O.O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC123840100
SDS View Documents
Thermo Scientific Chemicals
123840100
10 g Glass Bottle
Each for $124.11
Only null left
 
AC123841000
SDS View Documents
Thermo Scientific Chemicals
123841000
100 g Glass Bottle
Each for $888.40
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6153-64-6
496.469
IMLJLCJZQLGHJS-JEKSYDDFSA-N
(4S,4aR,5S,5aR,6S,12aR)-4-(dimethylamino)-1,5,6,10,11,12a-hexahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;dihydrate
C22H28N2O11
MFCD00151234
54680691
CC1(C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O)O.O.O

Specifications

Oxytetracycline Dihydrate
6153-64-6
Glass Bottle
4 to 8%
10 g
C22H28N2O11
14, 6976
CC1(C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O)O.O.O
496.469
496.46
Oxytetracycline dihydrate, 95%
Crystalline Powder
Authentic
On anhydrous substance: -203° to -216° (20°C, 589nm) (c=1, 0.1M HCl)
Yellow
95%
MFCD00151234
IMLJLCJZQLGHJS-JEKSYDDFSA-N
(4S,4aR,5S,5aR,6S,12aR)-4-(dimethylamino)-1,5,6,10,11,12a-hexahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;dihydrate
54680691
≥94% (on anhydrous substance)
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Suspected of damaging the unborn child.

GHS P Statement:
Use personal protective equipment as required.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LIGHT SENSITIVE

SDS
Documents

Documents

RUO – Research Use Only