missing translation for 'onlineSavingsMsg'
Learn More

Oxyresveratrol 96.0+%, TCI America™

Catalog No. p-7134301
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0373100MG 100 mg
O03731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0373100MG Supplier TCI America Supplier No. O0373100MG

Chemical Identifiers

CAS 29700-22-9
Molecular Formula C14H12O4
Molecular Weight (g/mol) 244.246
MDL Number MFCD11974969
InChI Key PDHAOJSHSJQANO-OWOJBTEDSA-N
Synonym 2,3′C,4,5′C-Tetrahydroxy-trans-stilbene
PubChem CID 5281717
ChEBI CHEBI:7870
IUPAC Name 4-[(E)-2-(3,5-dihydroxyphenyl)ethenyl]benzene-1,3-diol
SMILES C1=CC(=C(C=C1O)O)C=CC2=CC(=CC(=C2)O)O

Specifications

Melting Point 197°C
Color Yellow
Quantity 100 mg
Formula Weight 244.25
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Oxyresveratrol