missing translation for 'onlineSavingsMsg'
Learn More

Oxonic acid potassium salt

Catalog No. p-7041827
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6040006 5 g
AAJ6040014 25 g
AAJ6040022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6040006 Supplier Thermo Scientific Chemicals Supplier No. J6040006
Only null left

Inhibitor of uricase | CAS: 2207-75-2 | C4H2KN3O4 | 195.18 g/mol

Oxonic acid potassium salt is an inhibitor of uricase. This product has been used for the inhibition of 5-fluorouracil-induced gastrointestinal toxicity without the loss of its antitumor activity in rats. It has also been used to induce hyperuricemia in rats; as it inhibits uric acid metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxonic acid potassium salt is an inhibitor of uricase. This product has been used for the inhibition of 5-fluorouracil-induced gastrointestinal toxicity without the loss of its antitumor activity in rats. It has also been used to induce hyperuricemia in rats; as it inhibits uric acid metabolism.

Solubility
Soluble in hot water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 2207-75-2
Molecular Formula C4H2KN3O4
Molecular Weight (g/mol) 195.18
MDL Number MFCD00010565
InChI Key IAPCTXZQXAVYNG-UHFFFAOYSA-M
Synonym potassium oxonate, oteracil potassium, oxonic acid potassium salt, allantoxanic acid potassium salt, potassium azaorotate, oxonic acid, potassium salt, oxonate, potassium, oxonate, allantoxanic acid
PubChem CID 2723920
ChEBI CHEBI:80230
SMILES [K+].[O-]C(=O)C1=NC(=O)NC(=O)N1

Specifications

Melting Point 300°C
Color White
Physical Form Powder
Quantity 5 g
Merck Index 14,6947
Solubility Information Soluble in hot water.
IUPAC Name potassium;4,6-dioxo-1H-1,3,5-triazine-2-carboxylate
Formula Weight 195.18
Chemical Name or Material Oxonic acid potassium salt, 98%
EINECSNumber 218-627-5
RTECSNumber RR4580000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only