missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Yeast Autolysate Supplement

Catalog No. OXSR0105B
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR0105B 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXSR0105B Supplier Thermo Scientific™ Supplier No. SR0105B

Oxoid Yeast Autolysate Supplement is used for the preparation of New York City Medium for the isolation of Neisseria spp.

Oxoid Yeast Autolysate Supplement is used for the preparation of New York City Medium for the isolation of Neisseria spp.

  • Add to GC Agar Base, Part No. CM0367B
  • Each vial supplements 500mL of medium

Specifications

Description Yeast Autolysate Supplement
Form Freeze-dried
Product Type Microbiology Media
Quantity 10/Pk.
Yield Each vial for 500mL medium
Species Yeast