missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Vitox Supplement
Oxoid Vitox Supplement is used for the preparation of Thayer Martin Medium and New York City Medium for the isolation of Neisseria spp.
$400.93 - $1172.90
Specifications
| Description | Vitox Supplement |
|---|---|
| Form | Freeze-dried |
| Product Type | Microbiology Media |
| Packaging Type | 2 x 5/Pk. |
| Quantity | 5 x Freeze dried vial + 5 x Rehydration fluid |
Description
Oxoid Vitox Supplement is used for the preparation of Thayer Martin Medium and New York City Medium for the isolation of Neisseria spp.
- Contents: Vitox Supplement (5 vials) plus reconstitution fluid (5 vials)
- Add to GC Agar Base, Part No. CM0367B
Ordering Information:
- Each vial of Vitox plus reconstitution fluid supplements 500mL of medium
Includes:
- Five vials of Vitox plus five vials of reconstitution fluid
Specifications
| Vitox Supplement | |
| Microbiology Media | |
| 5 x Freeze dried vial + 5 x Rehydration fluid |
| Freeze-dried | |
| 2 x 5/Pk. |