missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Vitox Supplement

Catalog No. OXSR0090H
Change view
Click to view available options
Yield:
Each vial for 2L medium
Each vial for 500mL medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Yield
OXSR0090H Each vial for 2L medium
OXSR0090A Each vial for 500mL medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. OXSR0090H Supplier Thermo Scientific™ Supplier No. SR0090H

Oxoid Vitox Supplement is used for the preparation of Thayer Martin Medium and New York City Medium for the isolation of Neisseria spp.

Oxoid Vitox Supplement is used for the preparation of Thayer Martin Medium and New York City Medium for the isolation of Neisseria spp.

  • Contents: Vitox Supplement (5 vials) plus reconstitution fluid (5 vials)
  • Add to GC Agar Base, Part No. CM0367B

Ordering Information:

  • Each vial of Vitox plus reconstitution fluid supplements 500mL of medium

Includes:

  • Five vials of Vitox plus five vials of reconstitution fluid

Specifications

Description Vitox Supplement
Form Freeze-dried
Product Type Microbiology Media
Packaging Type 2 x 5/Pk.
Quantity 5 x Freeze dried vial + 5 x Rehydration fluid
Yield Each vial for 2L medium