missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Vegetable Peptone Broth

Catalog No. OXVG0101B
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXVG0101B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXVG0101B Supplier Thermo Scientific™ Supplier No. VG0101B

Thermo Scientific™ Oxoid Vegetable Peptone Broth is a meat-free general-purpose peptone.

Thermo Scientific™ Oxoid Vegetable Peptone Broth is a highly nutritious, general-purpose peptone.

An exceptional meat-free alternative to Typtone Soya Broth, it delivers equivalent performance in culture media or as a fermentation base.

  • Meat-free alternative to Typtone Soya Broth

Specifications

Content And Storage Dehydrated Powder, Ambient & Dry Storage
Form Dehydrated Powder
Product Type Peptone
For Use With (Application) Culture media or fermentation processes
Quantity 500 g
Protein Source Pea / Yeast

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.