Learn More
Thermo Scientific™ Oxoid™ VCNT Selective Supplement
Catalog No. SR0091H
Change view
Click to view available options
Yield:
Each vial for 2L medium
Catalog No. Yield
SR0091H Each vial for 2L medium
1 options
Catalog No. SR0091H Supplier Thermo Scientific™ Supplier No. SR0091H
Only null left
The Oxoid VCNT Selective Supplement is used for the preparation of Thayer Martin Medium for the isolation of pathogenic Neisseria spp.
Oxoid VCNT Selective Supplement is used for the preparation of Thayer Martin Medium for the isolation of pathogenic Neisseria spp.
  • Add to GC Agar Base, Part No. CM0367B
Specifications
Description VCNT Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 2L medium