missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ VCAT Selective Supplement

Catalog No. OXSR0104B
Change view
Click to view available options
Quantity:
500 mL of Base
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR0104B 500 mL of Base
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXSR0104B Supplier Thermo Scientific™ Supplier No. SR0104B

Used for the preparation of New York City Medium for the isolation of Neisseria gonorrhoeae.

Thermo Scientific™ Oxoid VCAT Selective Supplement is used for the preparation of New York City Medium for the isolation of Neisseria gonorrhoeae.

Add to GC Agar Base, Part No. CM0367B

Specifications

Description VCAT, 10 Vials, each supplements 500mL of base
Form Liquid
Product Type Selective Supplement
Quantity 500 mL of Base