missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ TTC Solution

Catalog No. OXSR0148A
Change view
Click to view available options
Quantity:
10 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR0148A 10 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXSR0148A Supplier Thermo Scientific™ Supplier No. SR0148A
Only null left

Oxoid TTC Solution (0.125%) is used for the enumeration and identification of coliform bacteria.

Oxoid TTC Solution (0.125%) is used for the enumeration and identification of coliform bacteria.

Add to Tergitol 7 Agar, Part No. CM0793B

Specifications

Description TTC Solution
Form Liquid
Product Type Solution
Quantity 10 x 2 mL
Yield Each vial for 100mL medium