missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Staph/Strep Selective Supplement

Catalog No. OXSR0070E
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR0070E 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXSR0070E Supplier Thermo Scientific™ Supplier No. SR0070E

Oxoid Staph/Strep Selective Supplement is used for preparation of Columbia CNA Agar.

Oxoid Staph/Strep Selective Supplement is used for preparation of Columbia CNA Agar.

  • Add to Columbia Blood Agar Base Part No. CM0331
  • Each vial supplements 500mL of medium

Specifications

Description Staph/Strep Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 500 g
Yield Each vial for 500mL medium
Species Staphylococcus species, Streptococcus species